The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001024616.1 | NP_001019787.2 | 9 - 10 | 1366 | 71 |
GenBank mRNA | AK045520.1 | - | 9 - 10 | 1388 | 71 |
AK081440.1 | - | 9 - 10 | 1376 | 71 | |
AK157020.1 | - | 9 - 10 | 1401 | 71 | |
AK162461.1 | - | 5 - 6 | 746 | 71 | |
AK171643.1 | - | 9 - 10 | 1377 | 71 | |
BC006855.1 | - | 1 - 2 | 3098 | 71 | |
BC020111.1 | - | 9 - 10 | 1374 | 71 | |
BC021476.1 | - | 9 - 10 | 1381 | 71 | |
BC023692.1 | - | 9 - 10 | 1394 | 71 | |
BC040791.1 | - | 1 - 2 | 3098 | 71 | |
BC051138.1 | - | 9 - 10 | 1380 | 71 | |
BC058585.1 | - | 9 - 10 | 1379 | 71 | |
BC085507.1 | - | 9 - 10 | 1366 | 71 |