The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001109657.3 | NP_001103127.1 | 13 - 14 | 1457 | 63 |
NM_001277079.1 | NP_001264008.1 | 13 - 14 | 1286 | 63 | |
NM_001277080.1 | NP_001264009.1 | 12 - 13 | 1200 | 63 | |
NM_008088.5 | NP_032114.2 | 13 - 14 | 1263 | 63 | |
GenBank mRNA | AF133184.1 | - | 9 - 10 | 1135 | 63 |
AK017394.1 | - | 13 - 14 | 1256 | 63 | |
AK076926.1 | - | 14 - 15 | 1340 | 63 | |
AK154463.1 | - | 13 - 14 | 1260 | 63 | |
AK172943.1 | - | 9 - 10 | 1079 | 63 | |
BC120528.1 | - | 13 - 14 | 1184 | 63 | |
BC120530.1 | - | 13 - 14 | 1184 | 63 | |
CV562268.1 | - | 5 - 6 | 501 | 63 | |
U19860.1 | - | 13 - 14 | 1289 | 63 |