The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_172584.3 | NP_766172.1 | 2 - 3 | 169 | 65 |
GenBank mRNA | AK038913.1 | - | 2 - 3 | 170 | 65 |
BC056464.1 | - | 2 - 3 | 150 | 65 |