The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001085407.1 | NP_001078876.1 | 8 - 9 | 1155 | 79 |
NM_001085408.1 | NP_001078877.1 | 7 - 8 | 1086 | 79 | |
NM_026563.3 | NP_080839.2 | 9 - 10 | 1305 | 79 | |
GenBank mRNA | AK014885.1 | - | 9 - 10 | 1274 | 79 |
AK016227.1 | - | 3 - 4 | 249 | 79 | |
AK082787.1 | - | 8 - 9 | 1126 | 79 | |
AK087227.1 | - | 3 - 4 | 671 | 79 | |
AK163690.1 | - | 7 - 8 | 1057 | 79 | |
BC005612.1 | - | 6 - 7 | 1429 | 79 | |
BC031199.1 | - | 7 - 8 | 1035 | 79 |