Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Gimap8
Entrez Gene ID
243374
Gene Name
GTPase, IMAP family member 8
Gene Aliases
Gm457, IAN9, IMAP8
Chromosome Location
Chr.6: 48647186 - 48660875 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001077410.1 NP_001070878.1 6 - 6 2261 114
NM_001286629.1 NP_001273558.1 5 - 5 1767 114
NM_212486.2 NP_997651.1 5 - 5 2255 114
GenBank mRNA AB178029.1 - 5 - 5 2255 114
AK134199.1 - 6 - 6 2262 114
AK155985.1 - 5 - 5 1768 114
BC137943.1 - 5 - 5 2232 114
BC137944.1 - 5 - 5 2232 114

View Full Product Details