Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ero1lb
Entrez Gene ID
67475
Gene Name
ERO1-like beta (S. cerevisiae)
Gene Aliases
1300013B24Rik, 1700065B09Rik, AI447560, ero1-beta
Chromosome Location
Chr.13: 12565809 - 12609530 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_026184.2 NP_080460.2 6 - 7 695 71
GenBank mRNA AF489856.1 - 6 - 7 501 71
AK004981.1 - - 672 71
AK006886.1 - 6 - 7 558 71
AK041951.1 - 6 - 7 671 71
AK160664.1 - 6 - 7 711 71
BC058721.1 - 6 - 7 695 71
BC111900.1 - 6 - 7 665 71
BC112426.1 - 6 - 7 665 71
BC145888.1 - 6 - 7 646 71
BC145890.1 - 6 - 7 646 71

View Full Product Details