Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Gp6
Entrez Gene ID
243816
Gene Name
glycoprotein 6 (platelet)
Gene Aliases
9830166G18Rik, Gm469, Gpvi
Chromosome Location
Chr.7: 4362853 - 4397905 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001163014.1 NP_001156486.1 2 - 3 129 117
GenBank mRNA AK079424.1 - 2 - 3 106 117
BC138170.1 - 2 - 3 113 117
BC145172.1 - 2 - 3 113 117

View Full Product Details