Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Lhx6
Entrez Gene ID
16874
Gene Name
LIM homeobox protein 6
Gene Aliases
Lhx6.1
Chromosome Location
Chr.2: 36081950 - 36105997 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001083125.1 NP_001076594.1 1 - 2 287 130
NM_008500.2 NP_032526.2 1 - 2 287 130
GenBank mRNA AK090339.2 - 1 - 2 135 130
AK140987.1 - 1 - 2 288 130

View Full Product Details