Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Npdc1
Entrez Gene ID
18146
Gene Name
neural proliferation, differentiation and control 1
Gene Aliases
AI314472, NPDC-1
Chromosome Location
Chr.2: 25403050 - 25409494 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008721.4 NP_032747.2 8 - 9 1110 68
GenBank mRNA AK146466.1 - 8 - 9 999 68
BC003320.1 - 8 - 9 1077 68
BC023887.1 - 7 - 8 1144 68
L03814.1 - 8 - 9 68
X67209.1 - 8 - 9 68

View Full Product Details