The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001085408.1 | NP_001078877.1 | 1 - 2 | 168 | 61 |
GenBank mRNA | AK163690.1 | - | 1 - 2 | 139 | 61 |
AW743523.1 | - | 1 - 2 | 118 | 61 | |
BC005612.1 | - | 1 - 2 | 159 | 61 | |
BC031199.1 | - | 1 - 2 | 117 | 61 |