The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001198872.1 | NP_001185801.1 | 15 - 16 | 1595 | 77 |
NM_001198873.1 | NP_001185802.1 | 15 - 16 | 1580 | 77 | |
NM_001198874.1 | NP_001185803.1 | 13 - 14 | 1526 | 77 | |
NM_001198875.1 | NP_001185804.1 | 13 - 14 | 1484 | 77 | |
NM_001198876.1 | NP_001185805.1 | 12 - 13 | 1466 | 77 | |
NM_001198877.1 | NP_001185806.1 | 15 - 16 | 1584 | 77 | |
NM_001198878.1 | NP_001185807.1 | 15 - 16 | 1569 | 77 | |
NM_010064.4 | NP_034194.1 | 12 - 13 | 1455 | 77 | |
GenBank mRNA | AF063231.1 | - | 12 - 13 | 1394 | 77 |
AK088015.1 | - | 12 - 13 | 1426 | 77 | |
AK141837.1 | - | 13 - 14 | 1494 | 77 | |
AK150173.1 | - | 12 - 13 | 1416 | 77 | |
AK161831.1 | - | 8 - 9 | 2116 | 77 | |
AK164322.1 | - | 13 - 14 | 1450 | 77 | |
AK164437.1 | - | 12 - 13 | 1390 | 77 | |
AK168730.1 | - | 12 - 13 | 1430 | 77 | |
BC139374.1 | - | 12 - 13 | 1350 | 77 | |
BC139375.1 | - | 12 - 13 | 1350 | 77 | |
GU992208.1 | - | 15 - 16 | 1486 | 77 | |
GU992209.1 | - | 15 - 16 | 1471 | 77 | |
GU992210.1 | - | 13 - 14 | 1375 | 77 | |
GU992211.1 | - | 15 - 16 | 1524 | 77 | |
GU992212.1 | - | 15 - 16 | 1509 | 77 |