The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_177190.5 | NP_796164.1 | 1 - 2 | 186 | 81 |
GenBank mRNA | AK082024.1 | - | 1 - 2 | 188 | 81 |