Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slc25a48
Entrez Gene ID
328258
Gene Name
solute carrier family 25, member 48
Gene Aliases
AU042651, E230025K15
Chromosome Location
Chr.13: 56437963 - 56472367 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_177809.4 NP_808477.2 2 - 3 231 70
GenBank mRNA AK054181.1 - 2 - 3 232 70
BC107250.1 - 2 - 3 195 70
BC107251.1 - 2 - 3 195 70

View Full Product Details