Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ammecr1
Entrez Gene ID
56068
Gene Name
Alport syndrome, mental retardation, midface hypoplasia and elliptocytosis chromosomal region gene 1
Gene Aliases
6230420G18Rik
Chromosome Location
Chr.X: 142853474 - 142966728 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_019496.3 NP_062369.1 2 - 3 636 93
GenBank mRNA AJ243485.1 - 2 - 3 636 93
AK051736.1 - 2 - 3 636 93

View Full Product Details