The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019496.3 | NP_062369.1 | 5 - 6 | 932 | 153 |
GenBank mRNA | AJ243485.1 | - | 5 - 6 | 932 | 153 |
AK051736.1 | - | 5 - 6 | 932 | 153 |