Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dnajc1
Entrez Gene ID
13418
Gene Name
DnaJ (Hsp40) homolog, subfamily C, member 1
Gene Aliases
4733401K02Rik, AA960110, D230036H06Rik, Dnajl1, ERdj1, ERj1p, MTJ1
Chromosome Location
Chr.2: 18206333 - 18392851 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001190817.1 NP_001177746.1 11 - 12 1920 78
NM_007869.3 NP_031895.1 11 - 12 1920 78
GenBank mRNA AK133582.1 - 11 - 12 1901 78
AK144228.1 - 11 - 12 1877 78
BC080300.1 - 11 - 12 1901 78
L16953.1 - 11 - 12 1597 78

View Full Product Details