Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Caprin1
Entrez Gene ID
53872
Gene Name
cell cycle associated protein 1
Gene Aliases
AL022980, Caprin-1, Gpiap, Gpiap1, Mmgpip137, P137, rng105
Chromosome Location
Chr.2: 103762945 - 103797640 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001111289.1 NP_001104759.1 16 - 17 2132 60
NM_001111290.1 NP_001104760.1 16 - 17 2105 60
NM_001111291.1 NP_001104761.1 17 - 18 2172 60
NM_001111292.1 NP_001104762.1 16 - 17 2132 60
NM_016739.3 NP_058019.2 17 - 18 2172 60
GenBank mRNA AB373955.1 - 16 - 17 1994 60
AK011150.1 - 2 - 3 161 60
AK029283.1 - 2 - 3 163 60
AK031050.1 - 2 - 3 163 60
AK141992.1 - 16 - 17 2074 60
AK147514.1 - 14 - 15 6214 60
AK149183.1 - 3 - 4 373 60
AK157707.1 - 17 - 18 2156 60
AK161131.1 - 11 - 12 1267 60
AK166461.1 - 16 - 17 2090 60
AK168676.1 - 16 - 17 2100 60
AK169366.1 - 8 - 9 903 60
AK170830.1 - 17 - 18 2134 60
BC052427.1 - 17 - 18 2148 60
BC127030.1 - 8 - 9 998 60
U18773.1 - 6 - 7 794 60
U27838.1 - 16 - 17 2109 60

View Full Product Details