The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001111289.1 | NP_001104759.1 | 1 - 2 | 348 | 77 |
NM_001111290.1 | NP_001104760.1 | 1 - 2 | 348 | 77 | |
NM_001111291.1 | NP_001104761.1 | 2 - 3 | 388 | 77 | |
NM_001111292.1 | NP_001104762.1 | 1 - 2 | 348 | 77 | |
NM_016739.3 | NP_058019.2 | 2 - 3 | 388 | 77 | |
GenBank mRNA | AB373955.1 | - | 1 - 2 | 210 | 77 |
AK133891.1 | - | 2 - 3 | 350 | 77 | |
AK141992.1 | - | 1 - 2 | 290 | 77 | |
AK147514.1 | - | 2 - 3 | 355 | 77 | |
AK148203.1 | - | 1 - 2 | 335 | 77 | |
AK157707.1 | - | 2 - 3 | 372 | 77 | |
AK166461.1 | - | 1 - 2 | 306 | 77 | |
AK168676.1 | - | 1 - 2 | 343 | 77 | |
AK170830.1 | - | 2 - 3 | 350 | 77 | |
BC052427.1 | - | 2 - 3 | 367 | 77 | |
CJ045742.1 | - | 2 - 3 | 389 | 77 | |
U27838.1 | - | 1 - 2 | 325 | 77 |