Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cenpi
Entrez Gene ID
102920
Gene Name
centromere protein I
Gene Aliases
CENP-I, Fshprh1
Chromosome Location
Chr.X: 134308089 - 134362643 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK043344.1 - 20 - 21 2290 76
AK145200.1 - 21 - 22 2346 76
AK161473.1 - 21 - 22 2429 76
BC024848.1 - 4 - 5 942 76
BC030328.1 - 21 - 22 2346 76

View Full Product Details