The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_145958.2 | NP_666070.2 | 2 - 3 | 812 | 69 |
GenBank mRNA | AK028396.1 | - | 2 - 3 | 758 | 69 |
AK129374.1 | - | 2 - 3 | 722 | 69 |