Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nipsnap3a
Entrez Gene ID
73398
Gene Name
nipsnap homolog 3A (C. elegans)
Gene Aliases
1700054F22Rik, AI429120
Chromosome Location
Chr.4: 52989284 - 53000854 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_028529.1 NP_082805.1 4 - 5 725 65

View Full Product Details