Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Aspn
Entrez Gene ID
66695
Gene Name
asporin
Gene Aliases
4631401G09Rik, AA986886, Plap1, Slrr1c
Chromosome Location
Chr.13: 49544443 - 49567565 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001172481.1 NP_001165952.1 2 - 3 537 157
NM_025711.3 NP_079987.2 2 - 3 554 157
GenBank mRNA AF316825.1 - 1 - 2 274 157
AK014504.1 - 2 - 3 510 157
AK085920.1 - 2 - 3 509 157
BC034888.1 - 2 - 3 456 157
BC145904.1 - 2 - 3 452 157
BC145930.1 - 2 - 3 435 157

View Full Product Details