The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_177867.3 | NP_808535.1 | 8 - 9 | 1572 | 89 |
GenBank mRNA | AK077155.1 | - | 8 - 9 | 1573 | 89 |
BC112427.1 | - | 7 - 8 | 1281 | 89 | |
BC113132.1 | - | 7 - 8 | 1281 | 89 |