The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001081168.1 | NP_001074637.1 | 1 - 2 | 655 | 88 |
GenBank mRNA | AK010234.1 | - | 1 - 2 | 143 | 88 |
BC051686.1 | - | 1 - 2 | 655 | 88 | |
BC094930.1 | - | 2 - 3 | 374 | 88 | |
BC117029.1 | - | 1 - 2 | 369 | 88 | |
BC117031.1 | - | 1 - 2 | 369 | 88 |