Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Hgd
Entrez Gene ID
15233
Gene Name
homogentisate 1, 2-dioxygenase
Gene Aliases
Hgo, aku
Chromosome Location
Chr.16: 37580153 - 37632026 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013547.3 NP_038575.2 4 - 5 456 68
NR_027778.1 3 - 4 367 68
GenBank mRNA BC024369.1 - 3 - 4 270 68
BC037628.1 - 4 - 5 370 68
BC044758.2 - 3 - 4 256 68
BC055029.1 - 4 - 5 337 68

View Full Product Details