Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Eif2b5
Entrez Gene ID
224045
Gene Name
eukaryotic translation initiation factor 2B, subunit 5 epsilon
Gene Aliases
C81315
Chromosome Location
Chr.16: 20498808 - 20509325 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172265.2 NP_758469.1 3 - 4 585 137
GenBank mRNA AK145801.1 - 3 - 4 587 137
AK160985.1 - 3 - 4 514 137
BC027268.1 - 3 - 4 531 138
BC038620.1 - 3 - 4 585 137
BC085255.1 - 3 - 4 496 137

View Full Product Details