Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gramd1c
Entrez Gene ID
207798
Gene Name
GRAM domain containing 1C
Gene Aliases
4921521N14Rik
Chromosome Location
Chr.16: 43980350 - 44065372 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001172107.1 NP_001165578.1 6 - 7 789 95
NM_153528.2 NP_705756.1 6 - 7 1019 95
GenBank mRNA AK029543.1 - 6 - 7 790 95
AK035257.1 - 6 - 7 1022 95
AK129016.1 - 6 - 7 958 95
BC037472.1 - 6 - 7 953 95

View Full Product Details