The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_028301.4 | NP_082577.2 | 14 - 15 | 1927 | 57 |
GenBank mRNA | AK012494.1 | - | 14 - 15 | 1876 | 57 |
AK129471.1 | - | 13 - 14 | 2956 | 57 | |
AK202800.1 | - | 1 - 2 | 55 | 57 | |
BC039644.1 | - | 7 - 8 | 880 | 57 | |
BC050929.1 | - | 14 - 15 | 1863 | 57 |