The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008576.3 | NP_032602.1 | 30 - 31 | 4513 | 70 |
GenBank mRNA | AK029876.1 | - | 30 - 31 | 4514 | 70 |
AK045109.1 | - | 2 - 3 | 315 | 70 | |
AK049174.1 | - | 14 - 15 | 2280 | 70 | |
AK140421.1 | - | 7 - 8 | 2056 | 70 | |
BC026509.1 | - | - | 440 | 70 | |
BC090617.1 | - | 30 - 31 | 4484 | 70 |