Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Hspbap1
Entrez Gene ID
66667
Gene Name
Hspb associated protein 1
Gene Aliases
3830421G21Rik
Chromosome Location
Chr.16: 35769663 - 35828462 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_175111.3 NP_780320.2 4 - 5 656 81
GenBank mRNA AK076189.1 - 4 - 5 623 81
AK089941.1 - 4 - 5 671 81

View Full Product Details