Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Gmip
Entrez Gene ID
78816
Gene Name
Gem-interacting protein
Gene Aliases
5031419I10Rik
Chromosome Location
Chr.8: 69808249 - 69821870 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_198101.1 NP_932769.1 9 - 10 913 98
GenBank mRNA AK052221.1 - 9 - 10 878 98
AK082337.1 - 9 - 10 910 98
AK137177.1 - 7 - 8 906 98
BC057037.1 - 9 - 10 913 98
BC060628.1 - 9 - 10 888 98

View Full Product Details