The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008223.3 | NP_032249.3 | 1 - 2 | 80 | 57 |
GenBank mRNA | BC089610.1 | - | 1 - 2 | 36 | 57 |
CA478051.1 | - | 1 - 2 | 117 | 57 |