Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ydjc
Entrez Gene ID
69101
Gene Name
YdjC homolog (bacterial)
Gene Aliases
1810015A11Rik, 4930521M19Rik
Chromosome Location
Chr.16: 17146882 - 17148857 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK076869.1 - - 319 94

View Full Product Details