The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_023243.5 | NP_075732.1 | 8 - 9 | 994 | 124 |
GenBank mRNA | AF287135.1 | - | 8 - 9 | 1047 | 124 |
AK007910.1 | - | 5 - 6 | 459 | 124 | |
AK078059.1 | - | 8 - 9 | 995 | 124 | |
BC038861.1 | - | 8 - 9 | 973 | 124 |