Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cyp4f17
Entrez Gene ID
208285
Gene Name
cytochrome P450, family 4, subfamily f, polypeptide 17
Gene Aliases
EG208285
Chromosome Location
Chr.17: 32506424 - 32529714 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001101445.1 NP_001094915.1 2 - 3 313 64

View Full Product Details