Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Fblim1
- Entrez Gene ID
-
74202
- Gene Name
- filamin binding LIM protein 1
- Gene Aliases
-
2410043F08Rik, Cal, Fblp1, Gt10, migfilin, migfilin(s)
- Chromosome Location
-
Chr.4: 141576062 - 141606052 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK010660.1 |
- |
1 - 2 |
149 |
85 |
View Full Product Details