Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cilp2
Entrez Gene ID
68709
Gene Name
cartilage intermediate layer protein 2
Gene Aliases
1110031K21Rik, AA407300, CLIP-2
Chromosome Location
Chr.8: 69880366 - 69887392 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_026818.2 NP_081094.2 1 - 2 106 71
GenBank mRNA BC006187.1 - 1 - 2 77 71
BC117911.2 - 7 - 8 71
BC117912.2 - 7 - 8 71

View Full Product Details