The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_026818.2 | NP_081094.2 | 7 - 8 | 1181 | 78 |
GenBank mRNA | BC006187.1 | - | 7 - 8 | 841 | 78 |
BC117911.2 | - | 1 - 2 | 78 | ||
BC117912.2 | - | 1 - 2 | 78 |