Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ssbp4
Entrez Gene ID
76900
Gene Name
single stranded DNA binding protein 4
Gene Aliases
1210002E11Rik, AW743380, Sspb4
Chromosome Location
Chr.8: 70597485 - 70608380 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_133772.2 NP_598533.2 6 - 7 560 160
GenBank mRNA AK154336.1 - 6 - 7 561 160
AK157387.1 - 6 - 7 549 162
BC002150.1 - 6 - 7 519 160
CX237617.1 - 5 - 6 342 160

View Full Product Details