Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ssbp4
Entrez Gene ID
76900
Gene Name
single stranded DNA binding protein 4
Gene Aliases
1210002E11Rik, AW743380, Sspb4
Chromosome Location
Chr.8: 70597485 - 70608380 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_133772.2 NP_598533.2 3 - 4 324 91
GenBank mRNA AK004835.1 - 3 - 4 436 91
AK154336.1 - 3 - 4 325 91
AK157387.1 - 3 - 4 313 91
BC002150.1 - 3 - 4 283 91
CX214457.1 - 3 - 4 324 91
CX237617.1 - 2 - 3 106 91

View Full Product Details