Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ccdc13
Entrez Gene ID
100502861
Gene Name
coiled-coil domain containing 13
Gene Aliases
2900041A11Rik, Gm188
Chromosome Location
Chr.9: 121797624 - 121839515 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_028384.1 NP_082660.1 11 - 12 1556 81
GenBank mRNA AK013632.1 - 1 - 2 73 81
AK038868.1 - 1 - 2 54 81
BB857029.1 - 1 - 2 130 81
BY296983.1 - 1 - 2 218 81

View Full Product Details