Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Catsper3
Entrez Gene ID
76856
Gene Name
cation channel, sperm associated 3
Gene Aliases
4921522D01Rik, CACRC
Chromosome Location
Chr.13: 55784579 - 55824942 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001252487.1 NP_001239416.1 5 - 6 897 68
NM_001252488.1 NP_001239417.1 5 - 6 744 68
NM_029772.4 NP_084048.1 5 - 6 858 68
GenBank mRNA AK014942.1 - 5 - 6 859 68
AY263400.1 - 5 - 6 872 68
BC089518.1 - - 997 68
HM179540.1 - 5 - 6 814 68
HM179541.1 - 5 - 6 661 68

View Full Product Details