The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_178397.3 | NP_848484.2 | 8 - 9 | 891 | 71 |
GenBank mRNA | AK122394.1 | - | 8 - 9 | 787 | 71 |
AK148529.1 | - | 8 - 9 | 852 | 71 | |
AK151617.1 | - | 8 - 9 | 796 | 71 | |
AK170111.1 | - | 8 - 9 | 875 | 71 | |
BC046817.1 | - | 8 - 9 | 853 | 71 | |
BC048767.1 | - | 7 - 8 | 699 | 71 |