Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Ice1
- Entrez Gene ID
-
218333
- Gene Name
- interactor of little elongation complex ELL subunit 1
- Gene Aliases
-
C77245, mKIAA0947
- Chromosome Location
-
Chr.13: 70588688 - 70637838 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC026613.1 |
- |
1 - 2 |
385 |
144 |
View Full Product Details