Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Lipi
Entrez Gene ID
320355
Gene Name
lipase, member I
Gene Aliases
D930038D03Rik, lpd1
Chromosome Location
Chr.16: 75540514 - 75586072 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001252513.1 NP_001239442.1 6 - 7 890 66
GenBank mRNA AK079027.1 - 4 - 5 301 66
AK086576.1 - 6 - 7 893 66
BC147403.1 - 1 - 2 82 66
BC147404.1 - 1 - 2 82 66

View Full Product Details