Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ccdc13
Entrez Gene ID
100502861
Gene Name
coiled-coil domain containing 13
Gene Aliases
2900041A11Rik, Gm188
Chromosome Location
Chr.9: 121797624 - 121839515 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_028384.1 NP_082660.1 9 - 10 1222 73
GenBank mRNA BM949737.1 - 2 - 3 310 73

View Full Product Details