Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Nr4a3
- Entrez Gene ID
-
18124
- Gene Name
- nuclear receptor subfamily 4, group A, member 3
- Gene Aliases
-
AI573420, CHN, CSMF, MINOR, NOR-1, Nor1, TEC
- Chromosome Location
-
Chr.4: 48044637 - 48086447 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC038520.1 |
- |
2 - 3 |
666 |
68 |
View Full Product Details