Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Olfr29-ps1
Entrez Gene ID
29848
Gene Name
olfactory receptor 29, pseudogene 1
Gene Aliases
MOR262-11, MTPCR52, Olfr158, Olfr29, Olfr37d, mOR37d
Chromosome Location
Chr.4: 43781365 - 43782327 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_033638.1 1 - 1 466 79
GenBank mRNA X89690.1 - 1 - 1 87 79

View Full Product Details