Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Eomes
Entrez Gene ID
13813
Gene Name
eomesodermin homolog (Xenopus laevis)
Gene Aliases
C77258, TBR-2, Tbr2
Chromosome Location
Chr.9: 118478180 - 118486132 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001164789.1 NP_001158261.1 3 - 4 1628 112
NM_010136.3 NP_034266.2 3 - 4 1628 112
GenBank mRNA AB031037.1 - 3 - 4 1600 112
AF013281.1 - 3 - 4 326 112
AK043327.1 - 3 - 4 959 112
AK089817.1 - 3 - 4 1188 112
AK143454.1 - 3 - 4 949 112
BC094319.1 - 3 - 4 1473 112

View Full Product Details