The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_144837.3 | NP_659086.2 | 13 - 14 | 5823 | 132 |
GenBank mRNA | AK129244.1 | - | 1 - 2 | 4260 | 132 |
AK134564.1 | - | 1 - 2 | 2266 | 132 |